2-Fluor-4-methoxybenzonitril

Lieferant: CHEMPUR

Synonyme: 2-Fluor-4-methoxybenzoesäurenitril

FL13834.POR
CHMPFL13834.POREA 0 EUR
CHMPFL13834.POR
2-Fluor-4-methoxybenzonitril
2-Fluor-4-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
MDL: MFCD00509374
CAS-Nummer: 94610-82-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR