4-Fluor-2-methoxybenzonitril

Lieferant: CHEMPUR

FL13837.POR
CHMPFL13837.POREA 0 EUR
CHMPFL13837.POR
4-Fluor-2-methoxybenzonitril
4-Fluor-2-methoxybenzonitril
Formel: C₈H₆FNO
MDL: MFCD04116335
CAS-Nummer: 191014-55-8
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR