3-Fluor-4-methoxybenzylalkohol

Lieferant: CHEMPUR

FL13838.POR
CHMPFL13838.POREA 0 EUR
CHMPFL13838.POR
3-Fluor-4-methoxybenzylalkohol
3-Fluor-4-methoxybenzylalkohol
Formel: C₈H₉FO₂
Molecular Weight: 156,16 g/mol
MDL: MFCD00070642
CAS-Nummer: 96047-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR