4-Fluor-3-methylphenylboronsäure

Lieferant: CHEMPUR

Synonyme: 3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid

FL13845.POR
CHMPFL13845.POREA 0 EUR
CHMPFL13845.POR
4-Fluor-3-methylphenylboronsäure
4-Fluor-3-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
MDL: MFCD01863527
CAS-Nummer: 139911-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR