3'-Fluor-4'-nitroacetanilid
Lieferant: CHEMPUR
Synonyme:
N-(3-fluoro-4-nitrophenyl)acetamide
FL13848.POR
CHMPFL13848.POREA
0
EUR
CHMPFL13848.POR
3'-Fluor-4'-nitroacetanilid
3'-Fluor-4'-nitroacetanilid
Formel:
C₈H₇FN₂O₃ Molecular Weight: 198,1512 g/mol |
MDL:
MFCD01463345 CAS-Nummer: 345-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...