4-Fluor-3-nitrobenzaldehyd

Lieferant: CHEMPUR

FL13849.POR 013849-25G 013849-5G 013849-1G
CHMPFL13849.POREA 0 EUR
CHMPFL13849.POR FLUO013849-25G FLUO013849-5G FLUO013849-1G
4-Fluor-3-nitrobenzaldehyd
4-Fluoro-3-nitrobenzaldehyde
Formel: C₇H₄FNO₃
Molecular Weight: 169.11 g/mol
MDL: MFCD01861388
CAS-Nummer: 42564-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR