2-Methoxy-5-(trifluormethoxy)benzoesäure

Lieferant: CHEMPUR

FL13861.POR
CHMPFL13861.POREA 0 EUR
CHMPFL13861.POR
2-Methoxy-5-(trifluormethoxy)benzoesäure
2-Methoxy-5-(trifluoromethoxy)benzoic acid
Formel: C₉H₇F₃O₄
MDL: MFCD01862055
CAS-Nummer: 191604-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR