4'-Fluor-3'-nitroacetophenon

Lieferant: CHEMPUR

FL13865.POR
CHMPFL13865.POREA 0 EUR
CHMPFL13865.POR
4'-Fluor-3'-nitroacetophenon
4'-Fluor-3'-nitroacetophenon
Formel: C₈H₆FNO₃
Molecular Weight: 183,14 g/mol
MDL: MFCD00115369
CAS-Nummer: 400-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR