2',3'-Dichloracetophenon
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
1-(2,3-Dichlorophenyl)ethanone
FL13876.POR
CHMPFL13876.POREA
0
EUR
CHMPFL13876.POR
2',3'-Dichloracetophenon
2',3'-Dichloracetophenon
Formel:
C₈H₆Cl₂O Molecular Weight: 189,04 g/mol |
MDL:
MFCD00052988 CAS-Nummer: 56041-57-7 EINECS: 259-954-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...