3-Methyl-2-thiophencarbonylchlorid

Lieferant: CHEMPUR

FL14753.1 014753-5G 014753-25G
CHMPFL14753.1EA 0 EUR
CHMPFL14753.1 FLUO014753-5G FLUO014753-25G
3-Methyl-2-thiophencarbonylchlorid
3-Methyl-2-thiophencarbonylchlorid
Formel: C₆H₅ClOS
MDL: MFCD00130091
CAS-Nummer: 61341-26-2
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR