4-tert-Butyl-2-methylthiazol

Lieferant: CHEMPUR

CFF873. FL17137.1
CHMPCFF873.EA 0 EUR
CHMPCFF873. CHMPFL17137.1
4-tert-Butyl-2-methylthiazol
4-tert-Butyl-2-methylthiazol
Formel: C₈H₁₃NS
Molecular Weight: 155,26 g/mol
MDL: MFCD00051643
CAS-Nummer: 15679-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR