ω-Brom-2,6-dihydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: α-Brom-2,6-dihydroxyacetophenon, 2-Brom-2',6'-dihydroxyacetophenon, 2-Brom-1-(2,6-dihydroxyphenyl)ethan-1-on, 2,6-Dihydroxyphenacylbromid

FL18102.POR
CHMPFL18102.POREA 0 EUR
CHMPFL18102.POR
ω-Brom-2,6-dihydroxyacetophenon
ω-Brom-2,6-dihydroxyacetophenon
Formel: C₈H₇BrO₃
CAS-Nummer: 2491-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR