ω-Brom-2,4-dihydroxyacetophenon
Lieferant: CHEMPUR
Synonyme:
2,4-Dihydroxyphenacylbromid, 2-Brom-1-(2,4-dihydroxyphenyl)ethan-1-on, 2-Brom-2',4'-dihydroxyacetophenon, α-Brom-2,4-dihydroxyacetophenon
FL18107.POR
CHMPFL18107.POREA
0
EUR
CHMPFL18107.POR
ω-Brom-2,4-dihydroxyacetophenon
ω-Brom-2,4-dihydroxyacetophenon
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol |
MDL:
MFCD03789098 CAS-Nummer: 2491-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...