ω-Brom-2,5-dihydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: 2-Brom-2',5'-dihydroxyacetophenon, 2-Brom-1-(2,5-dihydroxyphenyl)ethan-1-on, α-Brom-2,5-dihydroxyacetophenon, 2,5-Dihydroxyphenacylbromid

FL18108.POR
CHMPFL18108.POREA 0 EUR
CHMPFL18108.POR
ω-Brom-2,5-dihydroxyacetophenon
ω-Brom-2,5-dihydroxyacetophenon
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
CAS-Nummer: 25015-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR