ω-Brom-2,3-dihydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: α-Brom-2,3-dihydroxyacetophenon, 2-Brom-2',3'-dihydroxyacetophenon, 2-Brom-1-(2,3-dihydroxyphenyl)ethan-1-on, 2,3-Dihydroxyphenacylbromid

FL18109.POR
CHMPFL18109.POREA 0 EUR
CHMPFL18109.POR
ω-Brom-2,3-dihydroxyacetophenon
ω-Brom-2,3-dihydroxyacetophenon
Formel: C₈H₇BrO₃
CAS-Nummer: 19278-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR