ω-Brom-3,5-dihydroxyacetophenon
Lieferant: CHEMPUR
Synonyme:
2-Brom-3',5'-dihydroxyacetophenon, α-Brom-3,5-dihydroxyacetophenon, 3,5-Dihydroxyphenacylbromid, 2-Brom-1-(3,5-dihydroxyphenyl)ethan-1-on
FL18110.POR
CHMPFL18110.POREA
0
EUR
CHMPFL18110.POR
ω-Brom-3,5-dihydroxyacetophenon
ω-Brom-3,5-dihydroxyacetophenon
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol |
MDL:
MFCD03265442 CAS-Nummer: 62932-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...