ω-Brom-3,4-dihydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: 2-Brom-3',4'-dihydroxyacetophenon, 3,4-Dihydroxyphenacylbromid, 2-Brom-1-(3,4-dihydroxyphenyl)ethan-1-on, α-Brom-3,4-dihydroxyacetophenon

FL18111.POR
CHMPFL18111.POREA 0 EUR
CHMPFL18111.POR
ω-Brom-3,4-dihydroxyacetophenon
ω-Brom-3,4-dihydroxyacetophenon
Formel: C₈H₇BrO₃
CAS-Nummer: 40131-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR