4-Acetoxy-3-methoxybenzaldehyd

Lieferant: CHEMPUR

Synonyme: Vanillinacetat, 4-Formyl-2-methoxyphenylacetat

FL18149.10
CHMPFL18149.10EA 0 EUR
CHMPFL18149.10
4-Acetoxy-3-methoxybenzaldehyd
4-Acetoxy-3-methoxybenzaldehyd
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
MDL: MFCD00003362
CAS-Nummer: 881-68-5
EINECS: 212-920-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR