3-Brom-o-xylol
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
1-Brom-2,3-dimethylbenzol
FL18326.POR
CHMPFL18326.POREA
0
EUR
CHMPFL18326.POR
3-Brom-o-xylol
3-Brom-o-xylol
Formel:
C₈H₉Br Molecular Weight: 185.06 g/mol Siedepunkt: 214 Dichte: 1.365 g/cm³ (20 °C) |
MDL:
MFCD00000069 CAS-Nummer: 576-23-8 EINECS: 209-398-2 UN: 1701 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...