3,5-Dibrom-p-kresol

Lieferant: CHEMPUR

Synonyme: 3,5-Dibrom-4-methylphenol, 1,3-Dibrom-5-hydroxy-2-methylbenzol, 2,6-Dibrom-4-hydroxytoluol

FL18503.1
CHMPFL18503.1EA 0 EUR
CHMPFL18503.1
3,5-Dibrom-p-kresol
3,5-Dibrom-p-kresol
Formel: C₇H₆Br₂O
Molecular Weight: 265,93 g/mol
MDL: MFCD00013968
CAS-Nummer: 13979-81-2
EINECS: 237-763-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR