1H,1H,3H-Hexafluorbutylacrylat

Lieferant: CHEMPUR

FL19158.POR
CHMPFL19158.POREA 0 EUR
CHMPFL19158.POR
1H,1H,3H-Hexafluorbutylacrylat
1H,1H,3H-Hexafluorbutylacrylat
Formel: C₇H₆F₆O₂
MDL: MFCD00044091
CAS-Nummer: 54052-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR