5'-Ethyl-2'-methoxyacetophenon

Lieferant: CHEMPUR

FL19222.POR
CHMPFL19222.POREA 0 EUR
CHMPFL19222.POR
5'-Ethyl-2'-methoxyacetophenon
5'-Ethyl-2'-methoxyacetophenon
Formel: C₁₁H₁₄O₂
MDL: MFCD03789119
CAS-Nummer: 29643-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR