3',5'-Dichlor-2'-methoxyacetophenon

Lieferant: CHEMPUR

FL19223.POR
CHMPFL19223.POREA 0 EUR
CHMPFL19223.POR
3',5'-Dichlor-2'-methoxyacetophenon
3',5'-Dichlor-2'-methoxyacetophenon
Formel: C₉H₈Cl₂O₂
MDL: MFCD03789120
CAS-Nummer: 154260-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR