Indol-3-carbonitril

Lieferant: CHEMPUR

Synonyme: 3-Cyan-1H-indol, 3-Indolcarbonitril, 1H-Indol-3-carbonitril, 3-Cyanindol

FL19326.POR
CHMPFL19326.POREA 0 EUR
CHMPFL19326.POR
Indol-3-carbonitril
Indol-3-carbonitril
Formel: C₉H₆N₂
Molecular Weight: 142,16 g/mol
Siedepunkt: 350 °C (1013 hPa)
MDL: MFCD00022717
CAS-Nummer: 5457-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR