Tetrahydrothiophen
Lieferant: CHEMPUR
|
Gefahr
|
Synonyme:
THT
FL19455.POR
CHMPFL19455.POREA
0
EUR
CHMPFL19455.POR
Tetrahydrothiophen
Tetrahydrothiophen
Formel:
C₄H₈S Molecular Weight: 88,17 g/mol Siedepunkt: 119 °C (1013 hPa) Schmelzpunkt: –96 °C Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00005476 CAS-Nummer: 110-01-0 EINECS: 203-728-9 UN: 2412 ADR: 3,II Merck Index: 12,09357 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...