Cyclohexylisocyanid

Lieferant: CHEMPUR

FL21021.POR
CHMPFL21021.POREA 0 EUR
CHMPFL21021.POR
Cyclohexylisocyanid
Cyclohexylisocyanid
Formel: C₇H₁₁N
MDL: MFCD00003839
CAS-Nummer: 931-53-3
EINECS: 213-238-7
UN: 2929
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR