tert-Butylisocyanid

Lieferant: CHEMPUR

Synonyme: t-Butylisocyanide

FL21022.POR
CHMPFL21022.POREA 0 EUR
CHMPFL21022.POR
tert-Butylisocyanid
tert-Butylisocyanid
Formel: C₅H₉N
Molecular Weight: 83,13 g/mol
MDL: MFCD00000002
CAS-Nummer: 7188-38-7
EINECS: 230-553-5
UN: 1992
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR