n-Butylisocyanid

Lieferant: CHEMPUR

FL21027.POR
CHMPFL21027.POREA 0 EUR
CHMPFL21027.POR
n-Butylisocyanid
n-Butylisocyanid
Formel: C₅H₉N
Molecular Weight: 83,132 g/mol
MDL: MFCD00000008
CAS-Nummer: 2769-64-4
EINECS: 220-458-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR