Cyclopentylisocyanid

Lieferant: CHEMPUR

FL21030.POR
CHMPFL21030.POREA 0 EUR
CHMPFL21030.POR
Cyclopentylisocyanid
Cyclopentylisocyanid
Formel: C₆H₉N
MDL: MFCD02664624
CAS-Nummer: 68498-54-4
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR