4-Methoxybenzylisocyanid

Lieferant: CHEMPUR

Synonyme: 1-(Isocyanomethyl)-4-methoxybenzene

FL21037.POR 021037-1G
CHMPFL21037.POREA 0 EUR
CHMPFL21037.POR FLUO021037-1G
4-Methoxybenzylisocyanid
4-Methoxybenzylisocyanid
Formel: C₉H₉NO
Molecular Weight: 147.173 g/mol
MDL: MFCD02664572
CAS-Nummer: 1197-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR