3,5-Dimethylbenzylisocyanid

Lieferant: CHEMPUR

FL21045.POR
CHMPFL21045.POREA 0 EUR
CHMPFL21045.POR
3,5-Dimethylbenzylisocyanid
3,5-Dimethylbenzylisocyanid
Formel: C₁₀H₁₁N
CAS-Nummer: 612485-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR