3-Methoxybenzylisocyanid

Lieferant: CHEMPUR

FL21049.POR
CHMPFL21049.POREA 0 EUR
CHMPFL21049.POR
3-Methoxybenzylisocyanid
3-Methoxybenzylisocyanid
Formel: C₉H₉NO
CAS-Nummer: 177762-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR