2,6-Difluorbenzylisocyanid

Lieferant: CHEMPUR

FL21063.POR
CHMPFL21063.POREA 0 EUR
CHMPFL21063.POR
2,6-Difluorbenzylisocyanid
2,6-Difluorbenzylisocyanid
Formel: C₈H₅F₂N
CAS-Nummer: 730964-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR