4-Brombenzylisocyanid

Lieferant: CHEMPUR

FL21067.POR
CHMPFL21067.POREA 0 EUR
CHMPFL21067.POR
4-Brombenzylisocyanid
4-Brombenzylisocyanid
Formel: C₈H₆BrN
CAS-Nummer: 602261-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR