2-Chlor-6-fluorbenzylisocyanid

Lieferant: CHEMPUR

FL21068.POR
CHMPFL21068.POREA 0 EUR
CHMPFL21068.POR
2-Chlor-6-fluorbenzylisocyanid
2-Chlor-6-fluorbenzylisocyanid
Formel: C₈H₅ClFN
MDL: MFCD04117518
CAS-Nummer: 730964-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR