2-Fluorbenzylisocyanid

Lieferant: CHEMPUR

FL21070.POR
CHMPFL21070.POREA 0 EUR
CHMPFL21070.POR
2-Fluorbenzylisocyanid
2-Fluorbenzylisocyanid
Formel: C₈H₆FN
CAS-Nummer: 602261-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR