3-Fluorbenzylisocyanid

Lieferant: CHEMPUR

FL21074.POR
CHMPFL21074.POREA 0 EUR
CHMPFL21074.POR
3-Fluorbenzylisocyanid
3-Fluorbenzylisocyanid
Formel: C₈H₆FN
CAS-Nummer: 602261-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR