3-Nitrobenzylisocyanid

Lieferant: CHEMPUR

FL21075.POR
CHMPFL21075.POREA 0 EUR
CHMPFL21075.POR
3-Nitrobenzylisocyanid
3-Nitrobenzylisocyanid
Formel: C₈H₆N₂O₂
CAS-Nummer: 730964-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR