2,5-Difluorbenzylisocyanid

Lieferant: CHEMPUR

FL21080.POR
CHMPFL21080.POREA 0 EUR
CHMPFL21080.POR
2,5-Difluorbenzylisocyanid
2,5-Difluorbenzylisocyanid
Formel: C₈H₅F₂N
CAS-Nummer: 730964-56-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR