2,5-Dimethylbenzylisocyanid

Lieferant: CHEMPUR

FL21082.POR
CHMPFL21082.POREA 0 EUR
CHMPFL21082.POR
2,5-Dimethylbenzylisocyanid
2,5-Dimethylbenzylisocyanid
Formel: C₁₀H₁₁N
CAS-Nummer: 730964-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR