4-Methylbenzylisocyanid

Lieferant: CHEMPUR

Synonyme: 1-(Isocyanomethyl)-4-methylbenzene

FL21083.POR
CHMPFL21083.POREA 0 EUR
CHMPFL21083.POR
4-Methylbenzylisocyanid
4-Methylbenzylisocyanid
Formel: C₉H₉N
Molecular Weight: 131,174 g/mol
MDL: MFCD02664586
CAS-Nummer: 39495-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR