3-Methylbenzylisocyanid

Lieferant: CHEMPUR

FL21086.POR
CHMPFL21086.POREA 0 EUR
CHMPFL21086.POR
3-Methylbenzylisocyanid
3-Methylbenzylisocyanid
Formel: C₉H₉N
CAS-Nummer: 602261-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR