2-Methylbenzylisocyanid

Lieferant: CHEMPUR

FL21087.POR
CHMPFL21087.POREA 0 EUR
CHMPFL21087.POR
2-Methylbenzylisocyanid
2-Methylbenzylisocyanid
Formel: C₉H₉N
CAS-Nummer: 602261-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR