3,4-Dichlorbenzylisocyanid

Lieferant: CHEMPUR

FL21089.POR
CHMPFL21089.POREA 0 EUR
CHMPFL21089.POR
3,4-Dichlorbenzylisocyanid
3,4-Dichlorbenzylisocyanid
Formel: C₈H₅Cl₂N
CAS-Nummer: 1197-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR