2,4-Dichlorbenzylisocyanid

Lieferant: CHEMPUR

FL21090.POR
CHMPFL21090.POREA 0 EUR
CHMPFL21090.POR
2,4-Dichlorbenzylisocyanid
2,4-Dichlorbenzylisocyanid
Formel: C₈H₅Cl₂N
CAS-Nummer: 730964-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR