3'-Isocyanacetophenon

Lieferant: CHEMPUR

FL21179.POR
CHMPFL21179.POREA 0 EUR
CHMPFL21179.POR
3'-Isocyanacetophenon
3'-Isocyanacetophenon
Formel: C₉H₇NO
CAS-Nummer: 2980-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR