2-Isocyan-5-chlor-2'-fluorbenzophenon

Lieferant: CHEMPUR

FL21193.POR
CHMPFL21193.POREA 0 EUR
CHMPFL21193.POR
2-Isocyan-5-chlor-2'-fluorbenzophenon
2-Isocyan-5-chlor-2'-fluorbenzophenon
Formel: C₁₄H₇ClFNO
MDL: MFCD04117589
CAS-Nummer: 730964-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR