2-Isocyan-2'-fluor-5-brombenzophenon

Lieferant: CHEMPUR

FL21194.POR
CHMPFL21194.POREA 0 EUR
CHMPFL21194.POR
2-Isocyan-2'-fluor-5-brombenzophenon
2-Isocyan-2'-fluor-5-brombenzophenon
Formel: C₁₄H₇BrFNO
MDL: MFCD04117590
CAS-Nummer: 730964-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR