2-Isocyanbenzotrifluorid

Lieferant: CHEMPUR

FL21199.POR
CHMPFL21199.POREA 0 EUR
CHMPFL21199.POR
2-Isocyanbenzotrifluorid
2-Isocyanbenzotrifluorid
Formel: C₈H₄F₃N
MDL: MFCD02664547
CAS-Nummer: 105359-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR