3-Isocyanbenzotrifluorid

Lieferant: CHEMPUR

FL21207.POR
CHMPFL21207.POREA 0 EUR
CHMPFL21207.POR
3-Isocyanbenzotrifluorid
3-Isocyanbenzotrifluorid
Formel: C₈H₄F₃N
MDL: MFCD02664548
CAS-Nummer: 182276-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR